missing translation for 'onlineSavingsMsg'
Learn More

Dibromobimane, TRC

Product Code. 30384768
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384768 25 mg -
30378599 50 mg -
30374752 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384768 Supplier Toronto Research Chemicals Supplier No. TRCD42500025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68654-25-1
Molecular Formula C10 H10 Br2 N2 O2
Molecular Weight (g/mol) 350.01
IUPAC Name 1,7-bis(bromomethyl)-2,6-dimethylpyrazolo[1,2-a]pyrazole-3,5-dione
SMILES CC1=C(CBr)N2N(C(=O)C(=C2CBr)C)C1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H10Br2N2O2/c1-5-7(3-11)13-8(4-12)6(2)10(16)14(13)9(5)15/h3-4H2,1-2H3
Formula Weight 347.9109
Chemical Name or Material Dibromobimane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.