missing translation for 'onlineSavingsMsg'
Learn More

Dibromoacetonitrile, TRC

Product Code. 30378598
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378598 100 mg
30369438 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30378598 Supplier Toronto Research Chemicals Supplier No. TRCD422760100MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3252-43-5
Molecular Formula C2 H Br2 N
Molecular Weight (g/mol) 198.84
Synonym Acetonitrile, dibromo- (6CI,7CI,8CI,9CI), 2,2-Dibromoacetonitrile, Dibromoacetonitrile
IUPAC Name 2,2-dibromoacetonitrile
SMILES BrC(Br)C#N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C2HBr2N/c3-2(4)1-5/h2H
Formula Weight 196.8476
Chemical Name or Material Dibromoacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.