missing translation for 'onlineSavingsMsg'
Learn More

Dibromoacetonitrile, 94%

Product Code. 11468836
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468836 25 g 25g
11458836 5 g 5g
11478836 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468836 Supplier Thermo Scientific Alfa Aesar Supplier No. A16994.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3252-43-5 | C2HBr2N | 198.845 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3252-43-5
Molecular Formula C2HBr2N
Molecular Weight (g/mol) 198.845
MDL Number MFCD00001856
InChI Key NDSBDLSWTGLNQA-UHFFFAOYSA-N
Synonym dibromoacetonitrile, acetonitrile, dibromo, unii-vgj91h57xu, ccris 2673, 2,2-dibromoethanenitrile, vgj91h57xu, acetonitrile, 2,2-dibromo, dsstox_cid_4940, dsstox_rid_77590, dsstox_gsid_24940
PubChem CID 18617
ChEBI CHEBI:82445
IUPAC Name 2,2-dibromoacetonitrile
SMILES C(#N)C(Br)Br

Specifications

Boiling Point 67°C to 69°C (24 mmHg)
Flash Point 56°C (133°F)
Odor Pungent, Makes Eyes Water
Linear Formula Br2CHCN
Quantity 25 g
UN Number UN3286
Beilstein 1739037
Formula Weight 198.84
Percent Purity 94%
Chemical Name or Material Dibromoacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.