missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dibromo-p-xylene, 97%

Product Code. 10020810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10020810 100 g Glass bottle 100g
10520551 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10020810 Supplier Thermo Scientific Acros Supplier No. 112941000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-24-5 | C8H8Br2 | 263.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-24-5
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.95
MDL Number MFCD00000182
InChI Key RBZMSGOBSOCYHR-UHFFFAOYSA-N
Synonym 1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
PubChem CID 69335
IUPAC Name 1,4-bis(bromomethyl)benzene
SMILES C1=CC(=CC=C1CBr)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 142.0°C to 147.0°C
Color White to Beige
Density 2.0120g/mL
Boiling Point 245.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(CH2Br)2
Quantity 100 g
Beilstein 05, 385
Specific Gravity 2.012
Solubility Information Solubility in water: soluble,hydrolyses
Formula Weight 263.95
Percent Purity 97%
Physical Form Crystalline Powder and Coarse Crystals
Chemical Name or Material α,α′-Dibromo-p-xylene, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.