missing translation for 'onlineSavingsMsg'
Learn More

Dibromo(glyme)nickel, TRC

Product Code. 30381807
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381807 1 g
30378178 5 g
30386269 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381807 Supplier Toronto Research Chemicals Supplier No. TRCD4485081G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28923-39-9
Molecular Formula C4H10Br2NiO2
Molecular Weight (g/mol) 308.62
Synonym Nickel(II) Dimethoxyethane Bromide, Dibromo(1,2-dimethoxyethane)nickel(II), 1,2-Dimethoxyethane Nickel(II) Bromide, Dibromo[1,2-di(methoxy-kO)ethane]nickel, Dibromo(1,2-dimethoxyethane-O,O')nickel
SMILES [Ni+2].[Br-].[Br-].COCCOC

Specifications

Quantity 1 g
Formula Weight 305.8401
Chemical Name or Material Dibromo(glyme)nickel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.