missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diboc Biotin, TRC
Specifications
Specifications
| Chemical Name or Material | Diboc Biotin |
| Analyte or Component Names | Diboc Biotin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H32 N2 O7 S |
| Synonym | 5-((4S)-1,3-Bis(tert-butoxycarbonyl)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic Acid |
| InChI Formula | InChI=1S/C20H32N2O7S/c1-19(2,3)28-17(26)21-12-11-30-13(9-7-8-10-14(23)24)15(12)22(16(21)25)18(27)29-20(4,5)6/h12-13,15H,7-11H2,1-6H3,(H,23,24)/t12?,13-,15?/m0/s1 |
| Solution Type | Neat |
| SMILES | CC(C)(C)OC(=O)N1C2CS[C@@H](CCCCC(=O)O)C2N(C(=O)OC(C)(C)C)C1=O |
| IUPAC Name | 5-[(4S)-1,3-bis[(2-methylpropan-2-yl)oxycarbonyl]-2-oxo-3a,4,6,6a-tetrahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?