missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyldithiocarbamic Acid Zinc Salt, TRC

Product Code. 30380046
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380046 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380046 Supplier Toronto Research Chemicals Supplier No. TRCD4229931G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc Bis(dibenzyldithiocarbamate)
CAS 14726-36-4
Molecular Formula C30 H28 N2 S4 Zn
Quantity 1 g
Synonym Dibenzyldithiocarbamic Acid Zinc Salt, Zinc, bis[N,N-bis(phenylmethyl)carbamodithioato-κS,κS']-, (T-4)-, Zinc, bis(dibenzyldithiocarbamato)- (7CI,8CI), Zinc, bis[bis(phenylmethyl)carbamodithioato-S,S']-, (T-4)-, Zinc, bis[bis(phenylmethyl)carbamodithioato-κS,κS']-, (T-4)- (9CI), (T-4)-Bis[N,N-bis(phenylmethyl)carbamodithioato-κS,κS']zinc, (N,N-Dibenzyldithiocarbamato)zinc, Accelerator ZBEC, Arazate, Avorator ZBEC 70GE, Bis(dibenzyldithiocarbamato)zinc, DBZ, Nocceler Z, Nocceler ZTC, Perkacit ZBEC, Rhenogran ZBEC 70, Sanceler Z-BE, Vulkacit ZBEC, Vulkacit ZBEC/C, ZBEC, Zinc dibenzyldithiocarbamate, Zinc bis(dibenzyldithiocarbamate)
InChI Formula InChI=1S/2C15H15NS2.Zn/c2*17-15(18)16(11-13-7-3-1-4-8-13)12-14-9-5-2-6-10-14;/h2*1-10H,11-12H2,(H,17,18);/q;;+2/p-2
SMILES C(N(Cc1ccccc1)C2=[S][Zn+2]3([S-]C(=[S]3)N(Cc4ccccc4)Cc5ccccc5)[S-]2)c6ccccc6
Molecular Weight (g/mol) 610.226
Formula Weight 608.043
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.