missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyldithiocarbamic acid zinc salt, 95%

Product Code. 11985711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11985711 100 g Glass bottle 100g
10288580 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985711 Supplier Thermo Scientific Acros Supplier No. 406441000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14726-36-4 | C30H28N2S4Zn | 610.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14726-36-4
Molecular Formula C30H28N2S4Zn
Molecular Weight (g/mol) 610.21
InChI Key AUMBZPPBWALQRO-UHFFFAOYSA-L
Synonym zinc ii dibenzyldithiocarbamate, zinc dibenzyldithiocarbamate, dibenzyldithiocarbamic acid zinc salt, dibenzyldithiocarbamic acid, zinc salt, zinc n,n-dibenzylcarbamodithioate, zinc dibenzyl dithiocarbamate, zinc bis dibenzyldithiocarbamate, acmc-209cyb, ksc910q0r, bis dibenzyldithiocarbamic acid zinc
PubChem CID 84610
IUPAC Name zinc;N,N-dibenzylcarbamodithioate
SMILES C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].[Zn+2]

Spezifikation

Melting Point 183.0°C to 188.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Formula Weight 610.21
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Dibenzyldithiocarbamic acid zinc salt
compliance-icons
Produktidentifikator
  • Dibenzyldithiocarbamic acid zinc salt
Signalwort
  • Warning
Gefahrenkategorie
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Gefahrenhinweis
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Sicherheitshinweis
  • P273-Avoid release to the environment.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.