missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyldiselenide, 99+%, ACROS Organics™

Product Code. 10475423 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10475423 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10475423 Supplier Thermo Scientific Acros Supplier No. 293570010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1482-82-2
Melting Point 91°C to 94°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C14H14Se2
Linear Formula (C6H5CH2)2Se2
MDL Number MFCD00004767
Quantity 1 g
Beilstein 06, 470
Synonym dibenzyl diselenide, 1,2-dibenzyldiselane, benzyl diselenide, dibenzyldiselenide, dibenzyldiselane, diselenide, bis phenylmethyl, dibenzyldiselenium, bisbenzyldiselenide, diselenide,bis phenylmethyl, benzyldiselanyl methylbenzene
InChI Key HYAVEDMFTNAZQE-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C[Se][Se]CC2=CC=CC=C2
IUPAC Name (benzyldiselanyl)methylbenzene
Molecular Weight (g/mol) 340.208
PubChem CID 95955
Formula Weight 340.19
Percent Purity ≥99.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material Dibenzyldiselenide, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.