missing translation for 'onlineSavingsMsg'
Learn More

Dibenzylamine, 98%

Product Code. 10269213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269213 1 L Glass bottle 1 L
10234330 5 mL Glass bottle 5mL
10294370 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10269213 Supplier Thermo Scientific Acros Supplier No. 112610010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-49-1 | C14H15N | 197.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-49-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00004770
InChI Key BWLUMTFWVZZZND-UHFFFAOYSA-N
Synonym dibenzylamine, benzenemethanamine, n-phenylmethyl, n-benzylbenzylamine, bibenzylamine, dibenzyl-amine, dibenzyl amine, bisbenzylamine, n,n-dibenzylamine, n-benzylaminomethyl benzene, unii-3g0yfx01c6
PubChem CID 7656
SMILES C(NCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -26°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 300°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5CH2)2NH
Refractive Index 1.5740 to 1.576
Quantity 1 L
Beilstein 12, 1035
Merck Index 15, 3016
Specific Gravity 1.02
Solubility Information Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in alcohol,ether,toluene and chloroform,reacts with organic and inorganic acids
IUPAC Name N-benzyl-1-phenylmethanamine
Formula Weight 197.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dibenzylamine, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dibenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.