missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl succinate, 98%

Product Code. 11420808
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420808 25 g 25g
11430808 100 g 100g
11440808 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420808 Supplier Thermo Scientific Alfa Aesar Supplier No. B25257.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-43-5 | C18H18O4 | 298.34 g/mol

Dibenzyl succinate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl succinate is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-43-5
Molecular Formula C18H18O4
Molecular Weight (g/mol) 298.34
MDL Number MFCD00022034
InChI Key ODBOBZHTGBGYCK-UHFFFAOYSA-N
Synonym dibenzyl succinate, butanedioic acid, bis phenylmethyl ester, spasmine, benzyl succinate, dibenzylsuccinate, succinic acid dibenzyl ester, unii-ck14ug55av, ck14ug55av, diphenylmethyl butane-1,4-dioate, succinic acid, dibenzyl ester
PubChem CID 7653
ChEBI CHEBI:3058
IUPAC Name dibenzyl butanedioate
SMILES O=C(CCC(=O)OCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Melting Point 42°C to 47°C
Density 1.256
Boiling Point 245°C (15 mmHg)
Refractive Index 1.596
Quantity 25 g
Beilstein 1916056
Solubility Information Insoluble in water.
Formula Weight 298.34
Percent Purity 98%
Chemical Name or Material Dibenzyl succinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.