missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Phthalate, TRC

Product Code. 30376424
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376424 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376424 Supplier Toronto Research Chemicals Supplier No. TRCD4181501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-benzylester
CAS 523-31-9
Molecular Formula C22 H18 O4
Quantity 1 g
Synonym 1,2-Benzenedicarboxylic acid, 1,2-bis(phenylmethyl) ester, 1,2-Benzenedicarboxylic acid, bis(phenylmethyl) ester (9CI), Phthalic acid, dibenzyl ester (6CI,7CI,8CI), Benzyl phthalate, Dibenzyl phthalate, NSC 4057, Phthalic acid dibenzyl ester
InChI Formula InChI=1S/C22H18O4/c23-21(25-15-17-9-3-1-4-10-17)19-13-7-8-14-20(19)22(24)26-16-18-11-5-2-6-12-18/h1-14H,15-16H2
SMILES O=C(OCc1ccccc1)c2ccccc2C(=O)OCc3ccccc3
IUPAC Name dibenzyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 346.38
Formula Weight 346.1205
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.