missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Malonate, TRC

Product Code. 30354936
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30354936

Brand: Toronto Research Chemicals TRCD41788510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenzyl Malonate
CAS 15014-25-2
Molecular Formula C33 H30 N2 O4
Quantity 10 g
Synonym Propanedioic acid 1,3-bis(phenylmethyl) ester, Malonic acid, dibenzyl ester, Propanedioic acid bis(phenylmethyl) ester, Dibenzyl malonate, NSC 101027
InChI Formula InChI=1S/C33H30N2O4/c1-37-30-16-15-21(20-31(30)38-2)23-18-28-32(29(36)19-23)33(35-27-14-7-6-13-26(27)34-28)22-9-8-12-25(17-22)39-24-10-4-3-5-11-24/h3-17,20,23,33-35H,18-19H2,1-2H3
SMILES COc1ccc(cc1OC)C2CC3=C(C(Nc4ccccc4N3)c5cccc(Oc6ccccc6)c5)C(=O)C2
IUPAC Name 9-(3,4-dimethoxyphenyl)-6-(3-phenoxyphenyl)-5,6,8,9,10,11-hexahydrobenzo[b][1,4]benzodiazepin-7-one
Molecular Weight (g/mol) 518.602
Formula Weight 518.221
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.