missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Malonate, TRC

Product Code. 30358879
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358879 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358879 Supplier Toronto Research Chemicals Supplier No. TRCD41788550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenzyl Malonate
CAS 15014-25-2
Molecular Formula C33 H30 N2 O4
Quantity 50 g
Synonym Propanedioic acid 1,3-bis(phenylmethyl) ester, Malonic acid, dibenzyl ester, Propanedioic acid bis(phenylmethyl) ester, Dibenzyl malonate, NSC 101027
InChI Formula InChI=1S/C33H30N2O4/c1-37-30-16-15-21(20-31(30)38-2)23-18-28-32(29(36)19-23)33(35-27-14-7-6-13-26(27)34-28)22-9-8-12-25(17-22)39-24-10-4-3-5-11-24/h3-17,20,23,33-35H,18-19H2,1-2H3
SMILES COc1ccc(cc1OC)C2CC3=C(C(Nc4ccccc4N3)c5cccc(Oc6ccccc6)c5)C(=O)C2
IUPAC Name 9-(3,4-dimethoxyphenyl)-6-(3-phenoxyphenyl)-5,6,8,9,10,11-hexahydrobenzo[b][1,4]benzodiazepin-7-one
Molecular Weight (g/mol) 518.602
Formula Weight 518.221
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.