missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl malonate, 95%

Product Code. 11489812
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489812 50 g 50g
11479812 10 g 10g
11499812 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489812 Supplier Thermo Scientific Alfa Aesar Supplier No. A10844.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15014-25-2 | C17H16O4 | 284.311 g/mol

Dibenzyl malonate is used in the preparation of tetraethyl 3,3-bis(benzyloxycarbonyl)propylene bisphosphonate. It was also used in the preparation of benzyl umbelliferone-3-carboxylate via Knoevenagel condensation with 2,4-dihydroxybezaldehyde. It is a useful alternative to Diethyl­ malonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl malonate is used in the preparation of tetraethyl 3,3-bis(benzyloxycarbonyl)propylene bisphosphonate. It was also used in the preparation of benzyl umbelliferone-3-carboxylate via Knoevenagel condensation with 2,4-dihydroxybezaldehyde. It is a useful alternative to Diethyl­ malonate.

Solubility
Sparingly soluble in water; miscible in all proportions with ether and alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15014-25-2
Molecular Formula C17H16O4
Molecular Weight (g/mol) 284.311
MDL Number MFCD00004779
InChI Key RYFCSKVXWRJEOB-UHFFFAOYSA-N
Synonym dibenzyl malonate, propanedioic acid, bis phenylmethyl ester, 1,3-dibenzyl propanedioate, malonic acid dibenzyl ester, malonic acid dibenzyl, acmc-209d3u, diphenylmethyl propane-1,3-dioate, propanedioic acid,1,3-bis phenylmethyl ester, propanedioic acid, 1,3-bis phenylmethyl ester, s-3-amino-2-oxetanone p-toluenesulfonic acid salt boc protection form
PubChem CID 84754
IUPAC Name dibenzyl propanedioate
SMILES C1=CC=C(C=C1)COC(=O)CC(=O)OCC2=CC=CC=C2

Specifications

Density 1.137
Boiling Point 188°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.544
Quantity 50 g
Beilstein 1998264
Solubility Information Sparingly soluble in water; miscible in all proportions with ether and alcohol.
Formula Weight 284.31
Percent Purity 95%
Chemical Name or Material Dibenzyl malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.