missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl diselenide, 95%

Product Code. 11459177
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11459177

Brand: Thermo Scientific Alfa Aesar B21278.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1482-82-2 | C14H14Se2 | 340.208 g/mol

Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1482-82-2
Molecular Formula C14H14Se2
Molecular Weight (g/mol) 340.208
MDL Number MFCD00004767
InChI Key HYAVEDMFTNAZQE-UHFFFAOYSA-N
Synonym dibenzyl diselenide, 1,2-dibenzyldiselane, benzyl diselenide, dibenzyldiselenide, dibenzyldiselane, diselenide, bis phenylmethyl, dibenzyldiselenium, bisbenzyldiselenide, diselenide,bis phenylmethyl, benzyldiselanyl methylbenzene
PubChem CID 95955
IUPAC Name (benzyldiselanyl)methylbenzene
SMILES C1=CC=C(C=C1)C[Se][Se]CC2=CC=CC=C2

Specifications

Melting Point 91°C to 93°C
Quantity 25 g
UN Number UN3283
Beilstein 1877745
Solubility Information Soluble in water.
Formula Weight 340.18
Percent Purity 95%
Chemical Name or Material Dibenzyl diselenide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.