missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl diisopropylphosphoramidite, 90+%

Product Code. 11445237
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445237 25 g 25g
11435237 5 g 5g
11455237 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445237 Supplier Thermo Scientific Alfa Aesar Supplier No. B22629.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108549-23-1 | C20H28NO2P | 345.423 g/mol

Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Store under inert gas. Store at 0°C. Incompatible with oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108549-23-1
Molecular Formula C20H28NO2P
Molecular Weight (g/mol) 345.423
MDL Number MFCD00191988
InChI Key ANPWLBTUUNFQIO-UHFFFAOYSA-N
Synonym dibenzyl diisopropylphosphoramidite, dibenzyl n,n-diisopropylphosphoramidite, bis benzyloxy diisopropylamino phosphine, dibenzyldiisopropylphosphoramidite, dibenzyloxy diisopropylamino phosphine, bis benzyloxy phosphanyl diisopropylamine, phosphoramidous acid, bis 1-methylethyl-, bis phenylmethyl ester, n-dibenzyloxyphosphanyl-n-isopropyl-propan-2-amine, bis-bzl-2ipr-p, bis methylethyl bis phenylmethoxy phosphino amine
PubChem CID 196621
IUPAC Name N-bis(phenylmethoxy)phosphanyl-N-propan-2-ylpropan-2-amine
SMILES CC(C)N(C(C)C)P(OCC1=CC=CC=C1)OCC2=CC=CC=C2

Specifications

Density 1.03
Boiling Point 130°C (0.55 mmHg)
Flash Point 69°C (156°F)
Assay Percent Range 0.9
Quantity 25 g
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform.
Formula Weight 345.43
Percent Purity 95%
Chemical Name or Material Dibenzyl diisopropylphosphoramidite
compliance-icons
Product Identifier
  • Dibenzyl diisopropylphosphoramidite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Dibenzyl diisopropylphosphoramidite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.