missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl [1,3]Dioxole-4,4'-[1,3]dioxolan]-6-yl)methyl) Phosphate, TRC

Product Code. 30387978
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387978 5 mg 5mg
30384642 10 mg 10mg
30388746 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387978 Supplier Toronto Research Chemicals Supplier No. TRCD8396505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenzyl ((1,3)Dioxole-4,4'-((1,3)dioxolan-6-yl)methyl) Phosphate
Analyte or Component Names Dibenzyl ((1,3)Dioxole-4,4'-((1,3)dioxolan-6-yl)methyl) Phosphate
Shipping Condition Room Temperature
Molecular Formula C26 H33 O9 P
Synonym Dibenzyl (((3aS,4S,6R,6aS)-2,2,2',2'-Tetramethyldihydro-6H-spiro[furo[3,4-d][1,3]dioxole-4,4'-[1,3]dioxolan]-6-yl)methyl) Phosphate
InChI Formula InChI=1S/C26H33O9P/c1-24(2)28-18-26(35-24)23-22(33-25(3,4)34-23)21(32-26)17-31-36(27,29-15-19-11-7-5-8-12-19)30-16-20-13-9-6-10-14-20/h5-14,21-23H,15-18H2,1-4H3/t21-,22+,23+,26+/m1/s1
Solution Type Neat
SMILES CC1(C)O[C@H]2[C@@H](COP(=O)(OCc3ccccc3)OCc4ccccc4)O[C@]5(COC(C)(C)O5)[C@H]2O1
IUPAC Name [(3'aS,4S,6'R,6'aS)-2,2,2',2'-tetramethylspiro[1,3-dioxolane-4,4'-6,6a-dihydro-3aH-furo[3,4-d][1,3]dioxole]-6'-yl]methyl dibenzyl phosphate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 520.51
Quantity 5 mg
Formula Weight 520.186
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.