missing translation for 'onlineSavingsMsg'
Learn More

Dibenzoylmethane, TRC

Product Code. 30379242
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379242 25 g 25g
30381267 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379242 Supplier Toronto Research Chemicals Supplier No. TRCD42268325G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-46-7
Molecular Formula C15 H12 O2
Molecular Weight (g/mol) 224.25
Synonym 1,3-Diphenyl-1,3-propanedione, 1,3-Diphenylpropan-1,3-dione, 2-Benzoylacetophenone, AD 158, DBM, DBM 83, Dibenzoylmethane, Karenz DK 2, NSC 406806, NSC 52984, NSC 6266, PB 201, Phenyl phenacyl ketone, Rhodiastab 83, Rhodiastab 83P, ω-Benzoylacetophenone
IUPAC Name 1,3-diphenylpropane-1,3-dione
SMILES O=C(CC(=O)c1ccccc1)c2ccccc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C15H12O2/c16-14(12-7-3-1-4-8-12)11-15(17)13-9-5-2-6-10-13/h1-10H,11H2
Formula Weight 224.0837
Chemical Name or Material 2-Benzoylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.