missing translation for 'onlineSavingsMsg'
Learn More

Dibenzoyl peroxide, 97% (dry wt.), wet with 25% water

Product Code. 11337956
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337956 1000 g 1kg
11317956 50 g 50g
11327956 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337956 Supplier Thermo Scientific Alfa Aesar Supplier No. L13174.0B
  • ONLINE EXCLUSIVE PRICE
    £100.00
    £90.65 / 1kg
    Save £9.35
    9% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 94-36-0 | C14H10O4 | 242.23 g/mol

Benzoyl peroxide is widely utilized as a radical initiator to induce polymerizations. It finds applications for acne treatment, for bleaching flour, hair and teeth and for cross-linking polyester resins. It also has major applications in antiseptic and bleaching properties. It serves as a catalyst for polyester thermoset resins and as a hardener to start the polymerization process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoyl peroxide is widely utilized as a radical initiator to induce polymerizations. It finds applications for acne treatment, for bleaching flour, hair and teeth and for cross-linking polyester resins. It also has major applications in antiseptic and bleaching properties. It serves as a catalyst for polyester thermoset resins and as a hardener to start the polymerization process.

Solubility
Soluble in ether and chloroform. Slightlysoluble in ethanol. Insoluble in water.

Notes
Incompatible with strong oxidizer, reducing agents, acids, bases, alcohols and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-36-0
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00003071
InChI Key OMPJBNCRMGITSC-UHFFFAOYSA-N
Synonym benzoyl peroxide, dibenzoyl peroxide, peroxide, dibenzoyl, benzoperoxide, benzoyl superoxide, acetoxyl, lucidol, benoxyl, panoxyl, benzol peroxide
PubChem CID 7187
ChEBI CHEBI:82405
IUPAC Name benzoyl benzenecarboperoxoate
SMILES C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2

Specifications

Quantity 1000 g
UN Number UN3104
Beilstein 984320
Merck Index 14,1116
Solubility Information Soluble in ether and chloroform. Slightlysoluble in ethanol. Insoluble in water.
Formula Weight 242.23
Percent Purity 97%
Assay (dry wt.), wet with 25% water
Chemical Name or Material Benzoyl peroxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dibenzoyl peroxide, wet with 25% water
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Organic peroxide Type C & D
  • Skin sensitiser Category 1
Hazard Statement
  • H242-Heating may cause a fire.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Dibenzoyl peroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.