missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dibenzoyl-L-tartaric anhydride, 98%

Product Code. 10449291 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10449291 25 g Plastic bottle 25g
10439291 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10449291 Supplier Thermo Scientific Acros Supplier No. 336030250
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64339-95-3 | C18H12O7 | 340.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64339-95-3
Molecular Formula C18H12O7
Molecular Weight (g/mol) 340.29
MDL Number MFCD00067102
InChI Key OXIKRMSPXYQFOT-ZCWZLOQUNA-N
Synonym 3r,4r-2,5-dioxotetrahydrofuran-3,4-diyl dibenzoate, +-dibenzoyl-l-tartaric anhydride, 2,5-furandione, 3,4-bis benzoyloxy dihydro-, 3r,4r, 2-o,3-o-dibenzoyl-l-tartaric anhydride, 3r,4r-2,5-dioxo-3,4-dibenzoyloxy-3,4-dihydrofuran, 3r,4r-4-benzoyloxy-2,5-dioxooxolan-3-yl benzoate
PubChem CID 2733743
IUPAC Name [(3R,4R)-4-benzoyloxy-2,5-dioxooxolan-3-yl] benzoate
SMILES O=C(O[C@@H]1[C@@H](OC(=O)C2=CC=CC=C2)C(=O)OC1=O)C1=CC=CC=C1

Specifications

Melting Point 174°C
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ethyl ether
Formula Weight 340.29
Percent Purity 98%
Chemical Name or Material (+)-Dibenzoyl-L-tartaric anhydride
compliance-icons
Product Identifier
  • (+)-Dibenzoyl-L-tartaric anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.