missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-tartaric acid, 98%

Product Code. 10119889 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10119889 100 g Glass bottle 100g
10109889 25 g Glass bottle 25g
15119303 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10119889 Supplier Thermo Scientific Acros Supplier No. 438121000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2743-38-6 | C18H14O8 | 358.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2743-38-6
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
InChI Key YONLFQNRGZXBBF-ZIAGYGMSSA-N
Synonym dibenzoyl-l-tartaric acid, 2r,3r-2,3-bis benzoyloxy succinic acid, dibenzoyltartaric acid, --dibenzoyl-l-tartaric acid, dibenzoyl-l-tartaricacid, l-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r, unii-3q14b666vm, 2,3-bis benzoyloxy tartaric acid, o,o-dibenzoyl-+-tartaric acid
PubChem CID 75969
IUPAC Name (2R,3R)-2,3-dibenzoyloxybutanedioic acid
SMILES OC(=O)[C@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 150.0°C to 151.6°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Specific Rotation − 116.70
Formula Weight 358.31
Percent Purity 98%
Physical Form Crystalline Powder or Solid
Chemical Name or Material (-)-Dibenzoyl-L-tartaric acid
Show More Show Less
compliance-icons
Product Identifier
  • Dibenzoyl-L-tartaric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.