missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dibenzoyl-D-Tartaric Acid Monohydrate, TRC

Product Code. 30376229
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376229 2.5 g -
30375590 500 mg -
30372045 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376229 Supplier Toronto Research Chemicals Supplier No. TRCD4227982.5G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80822-15-7
Molecular Formula C18H14O8 . H2O
Molecular Weight (g/mol) 376.3
SMILES O.OC(=O)[C@@H](OC(=O)c1ccccc1)[C@H](OC(=O)c2ccccc2)C(=O)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C18H14O8.H2O/c19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12;/h1-10,13-14H,(H,19,20)(H,21,22);1H2/t13-,14-;/m0./s1
Formula Weight 376.0794
Chemical Name or Material (+)-Dibenzoyl-D-Tartaric Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.