Learn More
Dibenzothiophene, 98%, ACROS Organics™
Brand: Acros Organics 112540250
Code : 29
Additional Details : CAS Number : 132-65-0 Weight : 0.07500kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Packaging | Plastic bottle |
---|---|
Quantity | 25g |
Chemical Identifiers
132-65-0 | |
184.26 | |
IYYZUPMFVPLQIF-UHFFFAOYSA-N | |
3023 | |
dibenzothiophene |
C12H8S | |
MFCD00004969 | |
diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene | |
CHEBI:23681 | |
C1=CC=C2C(=C1)C3=CC=CC=C3S2 |
Specifications
Dibenzothiophene | |
Authentic | |
332°C to 333°C | |
97°C to 100°C | |
132-65-0 | |
C12H8S | |
17,72 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,benzene,chloroform | |
C1=CC=C2C(=C1)C3=CC=CC=C3S2 | |
184.26 | |
CHEBI:23681 | |
Crystalline Powder and/or Chunks |
170°C | |
Plastic bottle | |
Gray-Green to White | |
25g | |
97.5% min. (GC) | |
MFCD00004969 | |
diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene | |
IYYZUPMFVPLQIF-UHFFFAOYSA-N | |
dibenzothiophene | |
3023 | |
184.26 | |
98% |
Safety and Handling
- Dibenzothiophene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .