missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene, 98%

Product Code. 11475013
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475013 100 g 100g
11465013 25 g 25g
11485013 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475013 Supplier Thermo Scientific Alfa Aesar Supplier No. A12288.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 132-65-0 | C12H8S | 184.256 g/mol

Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-65-0
Molecular Formula C12H8S
Molecular Weight (g/mol) 184.256
MDL Number MFCD00004969
InChI Key IYYZUPMFVPLQIF-UHFFFAOYSA-N
Synonym diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene
PubChem CID 3023
ChEBI CHEBI:23681
IUPAC Name dibenzothiophene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3S2

Specifications

Melting Point 97°C to 100°C
Boiling Point 332°C to 333°C
Flash Point 170°C (338°F)
Quantity 100 g
UN Number UN3077
Beilstein 121101
Solubility Information Soluble in water.
Formula Weight 184.25
Percent Purity 98%
Chemical Name or Material Dibenzothiophene
compliance-icons compliance-icons
Product Identifier
  • Dibenzothiophene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Dibenzothiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.