missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene-5,5-dioxide, Thermo Scientific™

Product Code. 17585525 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17585525 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17585525 Supplier Thermo Scientific Acros Supplier No. 112550250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Alpha Vector DIBENZOTHIOPHENESULFONE
CAS 1016-05-3
Melting Point 231°C to 236°C
Color White to yellow
Infrared Spectrum Conforms
Packaging Glass Bottle
Molecular Formula C12H8O2S
MDL Number 00004970
Quantity 25 g
Beilstein 17-72
InChI Key IKJFYINYNJYDTA-UHFFFAOYSA-N
SMILES O=S1(=O)C2=CC=CC=C2C2=CC=CC=C12
IUPAC Name 8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene-8,8-dione
Molecular Weight (g/mol) 216.25
Physical Form Crystalline Powder
Chemical Name or Material Dibenzothiophene-5,5-dioxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.