missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene-2-ol, TRC

Product Code. 30180341
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180341 500 mg 500mg
30179760 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180341 Supplier Toronto Research Chemicals Supplier No. TRCD417345500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22439-65-2
Molecular Formula C12H8OS
Molecular Weight (g/mol) 200.26
Synonym 2-Hydroxydibenzothiophene, 3-Hydroxydiphenylenesulfide, Benzo[b]benzo[b]thiophen-2-ol;
SMILES Oc1ccc2sc3ccccc3c2c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H8OS/c13-8-5-6-12-10(7-8)9-3-1-2-4-11(9)14-12/h1-7,13H
Formula Weight 200.0296
Chemical Name or Material Dibenzothiophene-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.