missing translation for 'onlineSavingsMsg'
Learn More

Dibenzosuberol, TRC

Product Code. 30355010
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355010 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355010 Supplier Toronto Research Chemicals Supplier No. TRCD4173301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10,11-Dihydro-5H-dibenzo[a,d][7]-annulen-5-ol (Dibenzosuberol)
CAS 1210-34-0
Molecular Formula C15 H14 O
Quantity 1 g
Synonym 10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ol, Amitriptyline Hydrochloride Imp. G (EP), 10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ol, 5-Hydroxy-10,11-dihydro-5H-dibenzo[a,d]cycloheptene, Dibenzosuberol, NSC 86157
InChI Formula InChI=1S/C15H14O/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-8,15-16H,9-10H2
SMILES OC1c2ccccc2CCc3ccccc13
IUPAC Name 6,11-dihydro-5H-dibenzo[2,1-b:1',2'-e][7]annulen-11-ol
Molecular Weight (g/mol) 210.27
Formula Weight 210.1045
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.