missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran, 98%

Product Code. 11417847
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417847 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11417847 Supplier Thermo Scientific Alfa Aesar Supplier No. A16521.18
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 132-64-9 | C12H8O | 168.20 g/mol

Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.

Solubility
Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 132-64-9
Melting Point 78°C to 83°C
Density 1.3
Boiling Point 287°C to 289°C
Flash Point 130°C
Molecular Formula C12H8O
MDL Number MFCD00004968
Quantity 50 g
UN Number UN3077
Beilstein 121100
Synonym dibenzo b,d furan, diphenylene oxide, 2,2 '-biphenylene oxide, 2,2 '-biphenylylene oxide, dibenzofurans
Solubility Information Insoluble in wate(<1 mg/mL). Soluble in nonpolar organic solvents.
InChI Key TXCDCPKCNAJMEE-UHFFFAOYSA-N
SMILES O1C2=C(C=CC=C2)C2=C1C=CC=C2
IUPAC Name dibenzofuran
Molecular Weight (g/mol) 168.20
PubChem CID 568
ChEBI CHEBI:28145
Formula Weight 168.2
Percent Purity 98%
Chemical Name or Material Dibenzofuran
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.