missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran-4-boronic acid, 98+%

Product Code. 11389805
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389805 1 g 1g
11399805 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389805 Supplier Thermo Scientific Alfa Aesar Supplier No. L19830.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 100124-06-9 | C12H9BO3 | 212.011 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100124-06-9
Molecular Formula C12H9BO3
Molecular Weight (g/mol) 212.011
MDL Number MFCD00092336
InChI Key ZXHUJRZYLRVVNP-UHFFFAOYSA-N
Synonym dibenzofuran-4-boronic acid, dibenzo b,d furan-4-ylboronic acid, 4-dibenzofuranyl boronic acid, 4-dibenzofuranboronic acid, 4-dibenzofuranboronicacid, 4-dibenzofuranylboronic acid, boronic acid, 4-dibenzofuranyl, dibenzo b,d furan-4-boronic acid, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid
PubChem CID 2734328
IUPAC Name dibenzofuran-4-ylboronic acid
SMILES B(C1=C2C(=CC=C1)C3=CC=CC=C3O2)(O)O

Specifications

Melting Point 286°C to 291°C
Quantity 1 g
Beilstein 1645426
Formula Weight 212.01
Percent Purity ≥98%
Chemical Name or Material Dibenzofuran-4-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.