missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran-2-sulfonic acid hydrate, 97%, Thermo Scientific™

Product Code. 15461937
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461937 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15461937 Supplier Thermo Scientific Alfa Aesar Supplier No. 043899.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 83863-63-2
Melting Point 144°C to 145°C
Molecular Formula C12H7O4S
MDL Number MFCD00043577
Quantity 1 g
Sensitivity Hygroscopic
Synonym dibenzo b,d furan-2-sulfonic acid, 2-dibenzofuransulfonic acid, dibenzofuran-2-sulphonic acid, 2-dibenzofuransulfonicacid, dibenzofuran-2-sulfonsaure, janfyphfipgytq-uhfffaoysa, dibenzo b,d furan-2-sulfonicacid, dibenzo b,d furan-2-sulfonic acid #, 8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-sulfonic acid
InChI Key JANFYPHFIPGYTQ-UHFFFAOYSA-M
SMILES [O-]S(=O)(=O)C1=CC2=C(OC3=C2C=CC=C3)C=C1
IUPAC Name dibenzofuran-2-sulfonic acid
Molecular Weight (g/mol) 247.24
PubChem CID 522803
Formula Weight 248.24 (Anhydrous)
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Dibenzofuran-2-sulfonic acid hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.