missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzofuran-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Used as an pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as an pharmaceutical intermediate.
Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Used as an pharmaceutical intermediate.
Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 74°C |
Color | White |
Quantity | 250 mg |
Beilstein | 141044 |
Sensitivity | Air Sensitive |
IUPAC Name | dibenzofuran-2-carbaldehyde |
Formula Weight | 196.2 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Dibenzofuran-2-carboxaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'