missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[b,d]furan-2-carbaldehyde, ≥97%, Thermo Scientific™

Product Code. 10454153
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10454153 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10454153 Supplier Thermo Scientific Maybridge Supplier No. MO07102DA
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 5397-82-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C13H8O2
MDL Number MFCD00136272
Quantity 1 g
Synonym dibenzo b,d furan-2-carbaldehyde, 2-dibenzofurancarboxaldehyde, dibenzofuran-2-carboxaldehyde, dibenzo b,d furan-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde, 2-dibenzofurancarbaldehyde, dibenzo b furan-2-carbaldehyde, benzo b benzofuran-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene-4-carbaldehyde, 8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde
InChI Key OVJMIWIVPWPZMN-UHFFFAOYSA-N
SMILES O=CC1=CC2=C(OC3=C2C=CC=C3)C=C1
IUPAC Name 8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene-4-carbaldehyde
Molecular Weight (g/mol) 196.21
PubChem CID 220843
Formula Weight 196.2
Percent Purity 97%
Chemical Name or Material Dibenzo[b, d]furan-2-carbaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.