Learn More
Dibenzo-18-crown-6, 98+%
Description
Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to strong oxidizing agents, strong acids.
Specifications
Specifications
| Melting Point | 160°C to 164°C |
| Boiling Point | 380°C to 384°C (679 mmHg) |
| Quantity | 5 g |
| Beilstein | 1162153 |
| Merck Index | 14,2602 |
| Solubility Information | Sparingly soluble in water. |
| SMILES | C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1 |
| Molecular Weight (g/mol) | 360.406 |
| Formula Weight | 360.41 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
- Dibenzo-18-crown-6
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Dibenzo-18-crown-6
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.