missing translation for 'onlineSavingsMsg'
Learn More

Dibenzenechromium, C 69.2%, H 5.8%, Thermo Scientific Chemicals

Product Code. 11332658
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332658 0.5 g 0.5g
11342658 2 g 2g
11322658 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11332658 Supplier Thermo Scientific Alfa Aesar Supplier No. 024139.02

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-54-1
Molecular Formula C12H12Cr
Molecular Weight (g/mol) 208.224
MDL Number MFCD00015485
InChI Key HVURSIGIEONDKB-UHFFFAOYSA-N
Synonym bis benzene chromium, bis benzene chromium 0, chromium, bis benzene-8ci, dibenzolchrom, chromium, bis h6-benzene-9ci
PubChem CID 10932683
IUPAC Name benzene;chromium
SMILES C1=CC=CC=C1.C1=CC=CC=C1.[Cr]

Specifications

Melting Point 260°C to 265°C
Color Black
Linear Formula (C6H6)2Cr
Quantity 0.5 g
UN Number UN3391
Sensitivity Air & Moisture Sensitive
Formula Weight 208.22
Concentration or Composition (by Analyte or Components) C 69.2%, H 5.8%
Physical Form Crystalline
Chemical Name or Material Dibenzenechromium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.