missing translation for 'onlineSavingsMsg'
Learn More

Dibenz[a,j]acridine, TRC

Product Code. 30367989
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367989 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367989 Supplier Toronto Research Chemicals Supplier No. TRCD41691010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenz(A,J)Acridine
CAS 224-42-0
Purity Grade Notes HPLC
Molecular Formula C21 H13 N
Quantity 10 mg
Synonym Dibenz[a,j]acridine, 1,2:7,8-Dibenzacridine, 7-Azadibenz[a,j]anthracene, NSC 114903
InChI Formula InChI=1S/C21H13N/c1-3-7-16-14(5-1)9-11-20-18(16)13-19-17-8-4-2-6-15(17)10-12-21(19)22-20/h1-13H
SMILES c1ccc2c(c1)ccc3nc4ccc5ccccc5c4cc23
IUPAC Name Dibenz[a,j]acridine
Molecular Weight (g/mol) 279.33
Formula Weight 279.1048
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.