missing translation for 'onlineSavingsMsg'
Learn More

Diazoxide, TRC

Product Code. 30359631
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359631 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359631 Supplier Toronto Research Chemicals Supplier No. TRCD4173055G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diazoxide
CAS 364-98-7
Molecular Formula C8 H7 Cl N2 O2 S
Quantity 5 g
Synonym Diazoxide, 7-chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, 2H-1,2,4-Benzothiadiazine, 7-chloro-3-methyl-, 1,1-dioxide, 4H-1,2,4-Benzothiadiazine, 7-chloro-3-methyl-, 1,1-dioxide (7CI), 3-Methyl-7-chloro-1,2,4-benzothiadiazine 1,1-dioxide, 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, Diazoxide, Dizoxide, Eudemine injection, Hyperstat, Hypertonalum, Mutabase, NSC 64198, NSC 76130, Proglicem, Proglycem, SRG 95213, Sch 6783
InChI Formula InChI=1S/C8H7ClN2O2S/c1-5-10-7-3-2-6(9)4-8(7)14(12,13)11-5/h2-4H,1H3,(H,10,11)
SMILES CC1=Nc2ccc(Cl)cc2S(=O)(=O)N1
IUPAC Name 7-chloro-3-methyl-2H-1λ^{6},2,4-benzothiadiazine 1,1-dioxide
Molecular Weight (g/mol) 230.67
Formula Weight 229.9917
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.