missing translation for 'onlineSavingsMsg'
Learn More

Diazoxide, MedChemExpress

Product Code. 16468636
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468636 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468636 Supplier Medchem Express Supplier No. HYB1140100mg
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diazoxide (Sch-6783) is an ATP-sensitive potassium channel activator, has the potential for hyperinsulinism treatment.

Diazoxide (Sch-6783) is an ATP-sensitive potassium channel activator, has the potential for hyperinsulinism treatment.

Specifications

Chemical Name or Material Diazoxide
CAS 364-98-7
Color Grayish Brown
Purity Grade Notes Research
Molecular Formula C8H7ClN2O2S
Quantity 100 mg
Synonym Sch-6783 SRG-95213
Solubility Information DMSO : ≥ 35 mg/mL (151.73 mM)
SMILES CC(NC1=CC=C(Cl)C=C12)=NS2(=O)=O
Molecular Weight (g/mol) 230.67
Formula Weight 230.67
Percent Purity 98.48%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.