missing translation for 'onlineSavingsMsg'
Learn More

Diazolidinyl Urea (Technical Grade), TRC

Product Code. 30360924
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
Product Code. Quantity unitSize
30360924 25 g 25g
30351276 50 g 50g
2 options
This item is not returnable. View return policy

Product Code. 30360924

Brand: Toronto Research Chemicals TRCD41732325G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78491-02-8
Formula Weight 278.0862
InChI Formula InChI=1S/C8H14N4O7/c13-1-9-7(18)10(2-14)5-6(17)12(4-16)8(19)11(5)3-15/h5,13-16H,1-4H2,(H,9,18)
IUPAC Name 1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea
Molecular Formula C8H14N4O7
Molecular Weight (g/mol) 278.22
SMILES OCNC(=O)N(CO)C1N(CO)C(=O)N(CO)C1=O
Synonym N-[1,3-Bis(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]-N,N'-bis(hydroxymethyl)urea, Diazolidinylurea, Germaben II-E, Germall II, N-Hydroxymethyl-N-[1,3-di(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-N'-hydroxymethylurea
Chemical Name or Material Diazolidinyl Urea (Technical Grade)

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.