missing translation for 'onlineSavingsMsg'
Learn More

Diaza-18-crown-6, 98%

Product Code. 10432441 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
100mg
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432441 500 mg Glass bottle 500mg
10045773 100 mg Glass bottle 100mg
10035773 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10432441 Supplier Thermo Scientific Acros Supplier No. 275825000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23978-55-4 | C12H28N2O4 | 264.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23978-55-4
Molecular Formula C12H28N2O4
Molecular Weight (g/mol) 264.37
MDL Number MFCD00005112
InChI Key NLMDJJTUQPXZFG-UHFFFAOYSA-P
Synonym kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane
PubChem CID 72805
IUPAC Name 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
SMILES C1COCCOCC[NH2+]CCOCCOCC[NH2+]1

Specifications

Melting Point 111°C to 115°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,chloroform,acetone
Formula Weight 262.34
Percent Purity 98%
Physical Form Shiny Crystalline Powder
Chemical Name or Material Diaza-18-crown-6, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Diaza-18-crown-6
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.