missing translation for 'onlineSavingsMsg'
Learn More

Diatomaceous Earth, Non-washed, ∽90% SiO2, MP Biomedicals™

Product Code. 11431051 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431051 1 kg 1kg
11441051 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431051 Supplier MP Biomedicals Supplier No. 0215760691
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 61790-53-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Diatomaceous silica
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Physical Form Powder
Color Gray to White
Quantity 1 kg
Assay Percent Range ∽90%
Formula Weight 60.04
Chemical Name or Material Diatomaceous Earth, Non-washed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.