missing translation for 'onlineSavingsMsg'
Learn More

Diatomaceous earth, Calcined, Honeywell™ Fluka™

Product Code. 15667810 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15667810

Brand: Honeywell-Fluka D38771KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Powder, suitable for most filtrations

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Quantity 1 kg
Packaging Poly Bottle
UN Number NONH for all modes of transport
Chemical Name or Material Diatomaceous Earth, Calcined
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.