Learn More
Specac™ Diasqueeze Plus Kit with ZnSe lenses
Product Code. 15289937
Encompass_Preferred
Click to view available options
Diameter (Metric):
1.5 mm
Product Code. 15289937
Brand: Specac™ GS02556
A Universal Diamond Compression Cell designed for FTIR microscopes. Enables samples to be compressed to an ideal thickness for transmission experiments. Uses 2 type IIIa diamond windows mounted onto a hastelloy plate. Clear aperture of 1.5mm diameter. Also includes a Microfocus Beam Condenser with ZnSe lenses.
Specifications
Diameter (Metric) 1.5 mm
Includes Microfocus Beam Condenser with ZnSe lenses