missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIAPH3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14342-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the diaphanous subfamily of the formin family. Members of this family are involved in actin remodeling and regulate cell movement and adhesion. Mutations in this gene are associated with autosomal domit auditory neuropathy 1. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| DIAPH3 | |
| Polyclonal | |
| Unconjugated | |
| DIAPH3 | |
| diap3, DIAPH3, Diaphanous related formin 3, DRF3, mDia2, Protein diaphanous homolog 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 81624 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NSV4 | |
| DIAPH3 | |
| DIAPH3 Fusion Protein Ag5656 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction