missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIAPH3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545725
This item is not returnable.
View return policy
Description
Peptide sequence: ILEEVLEALT SAGEEKKIDR FFCIVEGLRH NSVQLQVACM QLINALVTSP.
This gene encodes a member of the diaphanous subfamily of the formin family. Members of this family are involved in actin remodeling and regulate cell movement and adhesion. Mutations in this gene are associated with autosomal dominant auditory neuropathy 1. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| DIAPH3 | |
| Polyclonal | |
| Unconjugated | |
| DIAPH3 | |
| 4930417P13Rik; AN; AUNA1; DIA2; diap3; Diaph3; diaphanous homolog 3; diaphanous homolog 3 (Drosophila); diaphanous related formin 3; diaphanous-related formin; diaphanous-related formin 3; diaphanous-related formin-3; DKFZp434C0931; DKFZp686A13178; Drf3; FLJ34705; mDia2; mKIAA4117; NSDAN; p134MDia2; Protein diaphanous homolog 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 81624 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NSV4 | |
| DIAPH3 | |
| Synthetic peptide directed towards the N-terminal of human DIAPH3 (aa 303-352). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction