missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIAPH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552173
This item is not returnable.
View return policy
Description
Immunogen sequence: GGSEEPGGGR SNKRSAGNRA ANEEETKNKP KLNIQIKTLA DDVRDRITSF RKSTVKKEKP LIQHPIDSQV AMSEFPAAQP LYDERSLNLS EKEVLDLFEK MMEDMNLNEE KKAPLRNKDF TTKREMVVQY ISAT Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 39%.
The product of this gene belongs to the diaphanous subfamily of the formin homology family of proteins. This gene may play a role in the development and normal function of the ovaries. Defects in this gene have been linked to premature ovarian failure 2. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| DIAPH2 | |
| Polyclonal | |
| Unconjugated | |
| DIAPH2 | |
| DIA; DIA2; Dia3; Diap2; Diaph2; diaphanous homolog 2; diaphanous homolog 2 (Drosophila); diaphanous related formin 2; diaphanous-related formin 2; diaphanous-related formin-2; diaphorase-2; DRF2; E430022I22Rik; FLJ11167; mDia3; mDia3 isoform; POF; POF2; Protein diaphanous homolog 2; RGD1564399; RP11-212B22.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1730, 317189, 54004 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60879, O70566 | |
| DIAPH2 | |
| Recombinant protein corresponding to Human DIAPH2. Recombinant protein control fragment (Product #RP-92145). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction