missing translation for 'onlineSavingsMsg'
Learn More

DIAPH1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18413694 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413694 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413694 Supplier Novus Biologicals Supplier No. NB600231JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DIAPH1 Polyclonal antibody specifically detects DIAPH1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIAPH1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DIA1, diaphanous homolog 1 (Drosophila), hDIA1, homolog) 1, LFHL1FLJ25265
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 1200 and the C-terminus (residue 1248) of human Diaphanous Homolog 1 using the numbering given in entry NP_005210.1 (GeneID 1729).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1729
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.