missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DIAPH1 Antibody [Allophycocyanin], Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NB600-230APC

Product Code. 30026891

  • £467.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DIAPH1 Polyclonal antibody specifically detects DIAPH1 in Human samples. It is validated for Immunoprecipitation,Western Blot,KnockDown
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIAPH1
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Immunoprecipitation, Western Blot, KnockDown
APC
DIA1, diaphanous homolog 1 (Drosophila), hDIA1, homolog) 1, LFHL1FLJ25265
Maps to a region between residues 1 and 50 of human Diaphanous Homolog 1 using the numbering given in entry NP_005210.1 (GeneID 1729).
0.1 mL
Signal Transduction
1729
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.