missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIAPH-2 Antibody (V78 P3C10-D3), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18260JF549
This item is not returnable.
View return policy
Description
DIAPH-2 Monoclonal antibody specifically detects DIAPH-2 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| DIAPH-2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| V78 P3C10-D3 | |
| Western Blot | |
| DIA2, DIAdiaphanous (Drosophila, homolog) 2, diaphanous homolog 2 (Drosophila), Diaphanous-related formin-2, diaphorase-2, DRF2, POF, POF2FLJ11167, protein diaphanous homolog 2 | |
| Synthetic peptide of Human DIAPH2 | |
| 0.1 mL | |
| Cell Cycle and Replication, Transcription Factors and Regulators | |
| 1730 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction