Learn More
DIAPH-2 Antibody (CL1113) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52967FR
Product Code. 30300668
  • £558.00 / 0.10mL
Description
DIAPH-2 Monoclonal antibody specifically detects DIAPH-2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIAPH-2
Monoclonal
DyLight 680
DIA2, DIAdiaphanous (Drosophila, homolog) 2, diaphanous homolog 2 (Drosophila), Diaphanous-related formin-2, diaphorase-2, DRF2, POF, POF2FLJ11167, protein diaphanous homolog 2
This antibody was developed using a recombinant protein derived from O60879, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Cycle and Replication, Transcription Factors and Regulators
1730
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL1113
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified